Skip to Content
Merck

Skip To

SML1928

UM-164 trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

2-[[6-[4-(2-Hydroxyethyl)-1-piperazinyl]-2-methyl-4-pyrimidinyl]amino]-N-[2-methyl-5-[[3-(trifluoromethyl)benzoyl]amino]phenyl]-5-thiazolecarboxamide trifluoroacetate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C30H31F3N8O3S · xC2HF3O2
CAS Number:
Molecular Weight:
640.68 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
To inquire about this product SML1928, please contact your local Merck office or dealer. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

OCCN(CC1)CCN1C2=CC(NC3=NC=C(C(NC4=CC(NC(C5=CC(C(F)(F)F)=CC=C5)=O)=CC=C4C)=O)S3)=NC(C)=N2

InChI

1S/C30H31F3N8O3S/c1-18-6-7-22(37-27(43)20-4-3-5-21(14-20)30(31,32)33)15-23(18)38-28(44)24-17-34-29(45-24)39-25-16-26(36-19(2)35-25)41-10-8-40(9-11-41)12-13-42/h3-7,14-17,42H,8-13H2,1-2H3,(H,37,43)(H,38,44)(H,34,35,36,39)

InChI key

ANEBQUSWQAQFQB-UHFFFAOYSA-N

Biochem/physiol Actions

UM-164 is a potent dual c-Src/p38 kinase inhibitor that exhibits potent anti-TNBC activity in xenograft models. UM-164 inhibits p38 kinase and binds to an inactive c-Src conformation. UM-164 has a high therapeutic index in vivo.
potent dual c-Src/p38 kinase inhibitor that exhibits potent anti-TNBC activity


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters