Skip to Content
Merck
All Photos(1)

Key Documents

SML1520

Sigma-Aldrich

GS39783

≥98% (HPLC)

Synonym(s):

GS-39783, N,N′-Dicyclopentyl-2-methylsulfanyl-5-nitro-pyrimidine-4,6-diamine, N4,N6-dicyclopentyl-2-(methylthio)-5-nitro-4,6-pyrimidinediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23N5O2S
CAS Number:
Molecular Weight:
337.44
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

color

faintly yellow to dark yellow

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

CSc1nc(NC2CCCC2)c(c(NC3CCCC3)n1)[N+]([O-])=O

InChI

1S/C15H23N5O2S/c1-23-15-18-13(16-10-6-2-3-7-10)12(20(21)22)14(19-15)17-11-8-4-5-9-11/h10-11H,2-9H2,1H3,(H2,16,17,18,19)

InChI key

GSGVDKOCBKBMGG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

GS39783 is a positive allosteric modulator (PAM) of GABA-B receptors. GABAB receptor positive allosteric modulators have potential for treatment of many neuro-psychiatric disorders as well as drug dependence and seizures. The receptor is a GPCR expressed throughout the CNS; it acts as both an auto-receptor and heteroreceptor to modulate GABA release and regulate K+ channels and Ca++ channels. GS39783 has been shown to inhibit alcohol self-administration and to have an antipsychotic-like effect in animal models of addiction and schizophrenia, respectively.
Positive allosteric modulator of GABAB receptors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service