Skip to Content
Merck

Skip To

SML1508

C-DIM12

≥98% (HPLC)

Synonym(s):

3,3′-[(4-Chlorophenyl)methylene]bis-1H-indole, 4-Chlorophenyl-3,3′-diindolylmethane, 4-Chlorophenyldi-3-indolylmethane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Please contact Customer Service for Availability
2 710,00 CZK
2 303,50 CZK
25 mg
Please contact Customer Service for Availability
11 000,00 CZK
9 350,00 CZK

About This Item

Empirical Formula (Hill Notation):
C23H17N2Cl
CAS Number:
Molecular Weight:
356.85
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
protect from light

2 303,50 CZK

List Price2 710,00 CZKSave 15%
Web-Only Promotion

Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

, light orange to dark red

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC(C=C1)=CC=C1C(C2=CNC3=C2C=CC=C3)C4=CNC5=C4C=CC=C5

InChI

1S/C23H17ClN2/c24-16-11-9-15(10-12-16)23(19-13-25-21-7-3-1-5-17(19)21)20-14-26-22-8-4-2-6-18(20)22/h1-14,23,25-26H

InChI key

LTLRXTDMXOFBDW-UHFFFAOYSA-N

Biochem/physiol Actions

C-DIM12 is a potent and selective nuclear receptor Nurr1 (NR4A2) activator that activates Nurr1 in urothelial carcinoma cells and in pancreatic cells. C-DIM12 suppresses inflammatory signaling in microglia.
C-DIM12 is a potent and selective nuclear receptor Nurr1 (NR4A2) activator.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1885SML2461L135
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

room temp

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

H2O: insoluble, DMSO: ≥20 mg/mL, ethanol: 24 mg/mL

storage condition

protect from light

storage condition

-

storage condition

-

storage condition

-


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters