Skip to Content
Merck
All Photos(1)

Key Documents

SML1447

Sigma-Aldrich

Clemizole hydrochloride

≥98% (HPLC)

Synonym(s):

1-(p-Chlorobenzyl)-2-(1-pyrrolidinylmethyl)benzimidazole hydrochloride, 1-(p-Chlorobenzyl)-2-pyrrolidylmethylenebenzimidazole hydrochloride, 1-[(4-Chlorophenyl)methyl]-2-(1-pyrrolidinylmethyl)-1H-benzimidazole hydrochloride, CID 2782 hydrochloride, NSC 46261 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CZK 3,550.00
25 MG
CZK 9,700.00

CZK 3,550.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
CZK 3,550.00
25 MG
CZK 9,700.00

About This Item

Empirical Formula (Hill Notation):
C19H20ClN3 · HCl
CAS Number:
Molecular Weight:
362.30
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

CZK 3,550.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

ClC(C=C1)=CC=C1CN2C(CN3CCCC3)=NC4=CC=CC=C42.[H]Cl

InChI

1S/C19H20ClN3.ClH/c20-16-9-7-15(8-10-16)13-23-18-6-2-1-5-17(18)21-19(23)14-22-11-3-4-12-22;/h1-2,5-10H,3-4,11-14H2;1H

InChI key

DNFMJYXRIMLMBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Clemizole is a potent anti-allergic drug.[1]

Biochem/physiol Actions

Clemizole is a potent and preferring inhibitor of TRPC5 that efficiently blocks heterologously expressed homomeric TRPC5 channels as well as heteromeric TRPC1:TRPC5 channels. Clemizole is a potent and selective H1 histamine receptor antagonist. Clemizole inhibited binding of HCV RNA by NS4B inhibit HCV RNA replication in cell culture.
Clemizole is a potent and preferring inhibitor of TRPC5; Antihistaminic.
Clemizole is an antihistamine and an inhibitor of NS4B (nonstructural protein 4B) RNA of HCV (hepatitis C virus). Studies in Scn1α (sodium channel, voltage-gated, type I, α) Zebrafish mutant show that this drug has the potential as a therapeutic agent in Dravet Syndrome treatment.[2]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott C Baraban et al.
Nature communications, 4, 2410-2410 (2013-09-05)
Dravet syndrome is a catastrophic pediatric epilepsy with severe intellectual disability, impaired social development and persistent drug-resistant seizures. One of its primary monogenic causes are mutations in Nav1.1 (SCN1A), a voltage-gated sodium channel. Here we characterize zebrafish Nav1.1 (scn1Lab) mutants
Clemizole hydrochloride.
Parvez M
Acta Crystallographica Section C, Structural Chemistry, 52(4), 904-905 (1996)
Alberto Arboit et al.
Cells, 9(2) (2020-02-09)
Persistent neural activity has been observed in vivo during working memory tasks, and supports short-term (up to tens of seconds) retention of information. While synaptic and intrinsic cellular mechanisms of persistent firing have been proposed, underlying cellular mechanisms are not
Caoimhin S Griffin et al.
Scientific reports, 8(1), 9264-9264 (2018-06-20)
Acetylcholine contracts the bladder by binding to muscarinic M3 receptors on the detrusor, leading to Ca2+ influx via voltage-gated Ca2+ channels. The cellular mechanisms linking these events are poorly understood, but studies have suggested that activation of TRPC4 channels could
Julia M Richter et al.
Molecular pharmacology, 86(5), 514-521 (2014-08-21)
Canonical transient receptor potential channel 5 (TRPC5) is a nonselective, Ca(2+)-permeable cation channel that belongs to the large family of transient receptor potential channels. It is predominantly found in the central nervous system with a high expression density in the

Questions

  1. In which solution clemizole will be dissolved ?

    1 answer
    1. As mentioned on the Product Specification Sheet, this product can be dissolved in warm water ? 2.0 mg/ml. The exact concentration will be listed on the lot specific Certificate of Analysis. A Certificate can be accessed in the DOCUMENTATION section under 'Certificate of Analysis'. Please see the link below:
      https://www.sigmaaldrich.com/product/sigma/sml1447#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service