Skip to Content
Merck
All Photos(1)

Documents

SML1434

Sigma-Aldrich

UNC2170 trifluoroacetate

≥98% (HPLC)

Synonym(s):

3-Bromo-N-(3-(tert-butylamino)propyl)benzamide trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21BrN2O · C2HF3O2
CAS Number:
Molecular Weight:
427.26
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

oil

color

colorless to yellow

storage temp.

2-8°C

SMILES string

BrC1=CC=CC(C(NCCCNC(C)(C)C)=O)=C1.FC(F)(C(O)=O)F

InChI

1S/C14H21BrN2O.C2HF3O2/c1-14(2,3)17-9-5-8-16-13(18)11-6-4-7-12(15)10-11;3-2(4,5)1(6)7/h4,6-7,10,17H,5,8-9H2,1-3H3,(H,16,18);(H,6,7)

InChI key

DPAGQICQYYQSQP-UHFFFAOYSA-N

Biochem/physiol Actions

UNC2170 is an inhibitor of DNA damage response protein 53BP1, which recognizes dimthyl lysine residues on lysine 20 on histone H4 that is involved in double-strand DNA breaks. 17-fold selectivity for 53BP1 as compared to other methyl-lysine (Kme) binding proteins tested.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service