Skip to Content
Merck

Skip To

SML1111

S 24795

≥98% (HPLC)

Synonym(s):

2-[2-(4-Bromophenyl)-2-oxoethyl]-1-methyl pyridinium iodide

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C14H13BrNO · I
CAS Number:
Molecular Weight:
418.07
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Assay:
≥98% (HPLC)
Form:
powder


assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

H2O: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

C[N+](C=CC=C1)=C1CC(C2=CC=C(Br)C=C2)=O.[I-]

InChI

1S/C14H13BrNO.HI/c1-16-9-3-2-4-13(16)10-14(17)11-5-7-12(15)8-6-11;/h2-9H,10H2,1H3;1H/q+1;/p-1

InChI key

LERUHBBUGAOYOD-UHFFFAOYSA-M

Biochem/physiol Actions

S 24795 is a selective partial agonist for the α7-subtype of nicotinic acetylcholine receptor (nAChR).
S 24795 is a selective partial agonist for the α7-subtype of nicotinic acetylcholine receptor (nAChR). S 24795 was shown to be effective at improving learning and memory in various animal models relevant to Alzheimer′s disease and age-related memory loss, and is used in investigating the roles of α7 nAChRs in aging, learning, memory, and cognition.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.


Still not finding the right product?

Explore all of our products under S 24795


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters