Skip to Content
Merck

Skip To

SML0961

Kobe0065

≥98% (HPLC)

Synonym(s):

N-(3-Chloro-4-methylphenyl)-2-[2,6-dinitro-4-(trifluoromethyl)phenyl]-hydrazinecarbothioamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Please contact Customer Service for Availability
2 030,00 CZK

About This Item

Empirical Formula (Hill Notation):
C15H11ClF3N5O4S
CAS Number:
Molecular Weight:
449.79
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

2 030,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(c(c(c1)[N+](=O)[O-])NNC(=S)Nc2cc(c(cc2)C)Cl)[N+](=O)[O-]

InChI

1S/C15H11ClF3N5O4S/c1-7-2-3-9(6-10(7)16)20-14(29)22-21-13-11(23(25)26)4-8(15(17,18)19)5-12(13)24(27)28/h2-6,21H,1H3,(H2,20,22,29)

InChI key

KSJVAYBCXSURMQ-UHFFFAOYSA-N

Biochem/physiol Actions

Kobe0065 binds to Ras-GTP, blocking interactions with Raf kinase. Kobe0065 inhibits anchorage-dependent and -independent growth, blocks MEK/ERK activity and induces apoptosis in H-rasG12V transformed NIH3T3 cells. The compound Kobe0065 also inhibts tumor growth in SW480 colon cancer cell xenografts.
Kobe0065 is an inhibitor of Ras activity.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1604SML2736SML0143
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 15 mg/mL, clear

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: ≥15 mg/mL

color

white to beige

color

white to beige

color

white to beige

color

white to tan


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters