Skip to Content
Merck

Skip To

SML0924

ML167

≥98% (HPLC)

Synonym(s):

5-[4-[[(5-Methyl-2-furanyl)methyl]amino]-6-quinazolinyl]-2-furanmethanol, CID 44968231, NCGC00188654

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Please contact Customer Service for Availability
2 810,00 CZK
2 388,50 CZK

About This Item

Empirical Formula (Hill Notation):
C19H17N3O3
CAS Number:
Molecular Weight:
335.36
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

2 388,50 CZK

List Price2 810,00 CZKSave 15%
Web-Only Promotion

Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

off-white to gray-brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

N(Cc4[o]c(cc4)C)c1ncnc2c1cc(cc2)c3[o]c(cc3)CO

InChI

1S/C19H17N3O3/c1-12-2-4-14(24-12)9-20-19-16-8-13(3-6-17(16)21-11-22-19)18-7-5-15(10-23)25-18/h2-8,11,23H,9-10H2,1H3,(H,20,21,22)

InChI key

ROCFOIBAEVAOLQ-UHFFFAOYSA-N

Biochem/physiol Actions

ML167 is a selective inhibitor of of Cdc2-like kinase 4 (Clk4).
ML167 is the first highly selective inhibitor of of Cdc2-like kinase 4 (Clk4) with an IC50 of 136 nM for Clk4, >10-fold selectivity for closely related kinases Clk1-3 and Dyrk1A/1B, and minimal activity against a panel of other kinases tested. The Cdc2-like kinases (Clk′s) and the dual-specificity tyrosine phosphorylation-regulated kinases (Dyrk′s) are involved in regulation of gene splicing. The Cdc2-like kinases phosphorylate the serine/arginine-rich (SR) proteins, a major component of the spliceosome.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1965SML1725SML2931
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 15 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

color

off-white to gray-brown

color

white to brown

color

yellow to orange

color

white to beige


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters