Skip to Content
Merck

Skip To

SML0912

SB 674042

≥98% (HPLC)

Synonym(s):

(2S)- 1-[[5-(2-Fluorophenyl)-2-methyl-4-thiazolyl]carbonyl]-2-[(5-phenyl-1,3,4-oxadiazol-2-yl)methyl]-pyrrolidine (9CI), (S)-1-[5-[2-Fluorophenyl]-2-methylthiazol-4-yl]-1-[2-[[5-phenyl-[1,3,4]oxadiazol-2-yl]methyl]pyrrolidin-1-yl]methanone, SB-674042

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Please contact Customer Service for Availability
12 600,00 CZK

About This Item

Empirical Formula (Hill Notation):
C24H21FN4O2S
CAS Number:
Molecular Weight:
448.51
UNSPSC Code:
51111800
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

12 600,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1c(cccc1)c2[s]c(nc2C(=O)N3[C@@H](CCC3)Cc4nnc([o]4)c5ccccc5)C

InChI

1S/C24H21FN4O2S/c1-15-26-21(22(32-15)18-11-5-6-12-19(18)25)24(30)29-13-7-10-17(29)14-20-27-28-23(31-20)16-8-3-2-4-9-16/h2-6,8-9,11-12,17H,7,10,13-14H2,1H3/t17-/m0/s1

InChI key

HYBZWVLPALMACV-KRWDZBQOSA-N

Biochem/physiol Actions

SB-674042 is potent and selective OX1R antagonist.
SB-674042 is potent and selective OX1R antagonist. SB-674042 binds with high affinity to OX1R and mediates intracellular calcium flux in OX1R-expressing cells.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2809SML2472SML2384
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

color

white to beige

color

white to beige

color

white to pink

color

white to beige


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters