Skip to Content

SML0901

ML095 hydrochloride

≥98% (HPLC)

Synonym(s):

1-(3,4-Dihydroxyphenyl)-2-(2-ethyl-1H-imidazol-1-yl)-Ethanone hydrochloride, CID-25067483 hydrochloride, MLS-0315848 hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H14N2O3 · HCl
CAS Number:
Molecular Weight:
282.72
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 5 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

ML095 is a specific inhibitor of placental alkaline phosphatase, with greater than 25-fold selectivity over tissue non-specific alkaline phosphatase (TNAP) and intestinal alkaline phosphatase (IAP). The IC50 valuess of ML095 against PLAP, TNAP and IAP are 2.1, >100 and 53 μM, respectively.
ML095 is a specific inhibitor of placental alkaline phosphatase.


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library