Skip to Content
Merck
All Photos(1)

Key Documents

SML0866

Sigma-Aldrich

BMS-191011

≥98% (HPLC)

Synonym(s):

3-[(5-Chloro-2-hydroxyphenyl)methyl]-5-[4-(trifluoromethyl)phenyl]-1,3,4-oxadiazol-2(3H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CZK 2,500.00
25 MG
CZK 9,780.00

CZK 2,500.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
CZK 2,500.00
25 MG
CZK 9,780.00

About This Item

Empirical Formula (Hill Notation):
C16H10ClF3N2O3
CAS Number:
Molecular Weight:
370.71
UNSPSC Code:
12352200
NACRES:
NA.77

CZK 2,500.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

FC(F)(F)c1ccc(cc1)C2=NN(C(=O)O2)Cc3c(ccc(c3)Cl)O

InChI

1S/C16H10ClF3N2O3/c17-12-5-6-13(23)10(7-12)8-22-15(24)25-14(21-22)9-1-3-11(4-2-9)16(18,19)20/h1-7,23H,8H2

InChI key

QKOWACXSXTXRKA-UHFFFAOYSA-N

Biochem/physiol Actions

BMS-191011 is a potent opener of BKCa opener (large-conductance Ca2+-activated K channel).
BMS-191011 is a potent opener of BKCa opener (large-conductance Ca2+-activated K channel). The compound BMS-191011 has neuroprotective properties in rodent models of stroke, and induces dialation of rat retinal arterioles.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to potassium channels for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service