Skip to Content
Merck

Skip To

SML0716

SB225002

≥98% (HPLC), chemokine receptor CXCR2 inhibitor, powder

Synonym(s):

N-(2-Hydroxy-4-nitrophenyl)-N′-(2-bromophenyl)urea, SB-225002

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Please contact Customer Service for Availability
2 980,00 CZK
25 mg
Please contact Customer Service for Availability
12 000,00 CZK

About This Item

Empirical Formula (Hill Notation):
C13H10BrN3O4
CAS Number:
Molecular Weight:
352.14
UNSPSC Code:
51111800
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

2 980,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

SB225002, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

Brc1c(cccc1)NC(=O)Nc2c(cc(cc2)[N+](=O)[O-])O

InChI

1S/C13H10BrN3O4/c14-9-3-1-2-4-10(9)15-13(19)16-11-6-5-8(17(20)21)7-12(11)18/h1-7,18H,(H2,15,16,19)

InChI key

MQBZVUNNWUIPMK-UHFFFAOYSA-N

Biochem/physiol Actions

SB-225002 is a potent nonpeptide inhibitor of chemokine receptor CXCR2 with an IC50 of 22 nM for inhibiting interleukin IL-8 binding to CXCR2 and > 150-fold selectivity over CXCR1 receptors. CXCR2 binds many different immune cell chemoattractants. SB-225002 is crucial for cancer progression and is involved in inflammatory diseases like COPD, rheumatoid arthritis, and ulcerative colitis.
SB-225002 is a potent nonpeptide inhibitor of chemokine receptor CXCR2.
Blocking action of SB-225002 on CXCR2 (interleukin 8 receptor β), prevents the IL-8 induced matrix metalloproteinase-9 release from neutrophils. Matrix metalloproteinase-9 are proteolytic enzymes associated with inflammation.

Features and Benefits

This compound is featured on the Chemokine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0421SML1530SML0220
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

room temp

storage temp.

2-8°C

storage temp.

room temp

storage temp.

room temp

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 15 mg/mL, clear

solubility

DMSO: 5 mg/mL, clear (warmed)

solubility

DMSO: 5 mg/mL (clear solution)

color

white to beige

color

white to light brown

color

white to beige

color

white to beige


Still not finding the right product?

Explore all of our products under SB225002


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters