Skip to Content
Merck

Skip To

SML0607

Ceranib-2

≥98% (HPLC)

Synonym(s):

3-[3-(4-Methoxyphenyl)-1-oxo-2-propen-1-yl]-4-phenyl-2(1H)-quinolinone, 3-[3-(4-Methoxyphenyl)-1-oxo-2-propenyl]-4-phenyl-2(1H)-quinolinone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Please contact Customer Service for Availability
1 990,00 CZK

About This Item

Empirical Formula (Hill Notation):
C25H19NO3
CAS Number:
Molecular Weight:
381.42
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder

1 990,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

Ceranib-2 is a nonlipid ceramidase inhibitor that inhibits cellular ceramidase activity, decreases levels of sphingosine and sphingosine-1-phosphate (S1P), and inhibits cell proliferation.
Ceranib-2 is a nonlipid ceramidase inhibitor.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2689SML1085L2545
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 10 mg/mL, clear (warmed)

solubility

DMSO: >20 mg/mL

shipped in

wet ice

shipped in

-

shipped in

-

shipped in

-


Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters