Skip to Content
Merck
All Photos(1)

Documents

SML0487

Sigma-Aldrich

CID 2818500

≥98% (HPLC)

Synonym(s):

(Z)-α-Nitrostilbene, 1,1′-[(1Z)-1-Nitro-1,2-ethenediyl]bis-benzene, trans-α-Nitrostilbene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11NO2
CAS Number:
Molecular Weight:
225.24
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

Biochem/physiol Actions

CID 2818500 (trans-α-Nitrostilbene; 1,1′-[(1Z)-1-Nitro-1,2-ethenediyl]bis-benzene) is a selective inhibitor of Protein Arginine Methyltransferases PRMT1 and PRMT8. PRMT1 is responsible for the majority of arginine methylation in the cell involved in transcription, chromatin modification, and signal transduction. CID 2818500 has an IC50 values of 11 μM for PRMT1, and does not inhibit PRMTs 3, 4, and 6, although it does inhibit PRMT8. The mechanism is believed to involve a specific S-adenosylmethionine (SAM)-binding cysteine present only in the Type I PRMTs PRMT1 and PRMT8 and absent in other SAM-dependent methyltransferases.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service