Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
5 mg | Please contact Customer Service for Availability | 3 270,00 CZK |
25 mg | Please contact Customer Service for Availability | 13 000,00 CZK 11 050,00 CZK |
About This Item
Empirical Formula (Hill Notation):
C7H18NO2P
CAS Number:
Molecular Weight:
179.20
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥97% (NMR)
Form:
powder
Storage condition:
desiccated
Quality Segment
assay
≥97% (NMR)
form
powder
storage condition
desiccated
color
white to beige
solubility
H2O: 10 mg/mL (clear solution, warmed)
storage temp.
room temp
SMILES string
CCCCP(O)(=O)CCCN
InChI
1S/C7H18NO2P/c1-2-3-6-11(9,10)7-4-5-8/h2-8H2,1H3,(H,9,10)
InChI key
ONNMDRQRSGKZCN-UHFFFAOYSA-N
Biochem/physiol Actions
CGP36742 is a GABAB receptor antagonist.
CGP36742 was the first orally active GABAB-selective antagonist. The IC50 for blocking GABAergic transmission is 36 μM. Oral administration of CGP36742 in rats blocks baclofen-induced neuronal depression, and reduces learning deficit and passive avoidance in an olfactory bulbectomy depression model.
Features and Benefits
This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
1 of 1
This Item | |||
|---|---|---|---|
| description ≥97% (NMR) | description ≥97% (NMR), solid | description ≥98% (HPLC) | description ≥98% (HPLC), powder |
| form powder | form solid | form powder | form powder |
| assay ≥97% (NMR) | assay ≥97% (NMR) | assay ≥98% (HPLC) | assay ≥98% (HPLC) |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| storage temp. room temp | storage temp. −20°C | storage temp. room temp | storage temp. room temp |
| solubility H2O: 10 mg/mL (clear solution, warmed) | solubility H2O: >20 mg/mL | solubility DMSO: 5 mg/mL, clear (warmed) | solubility DMSO: 10 mg/mL, clear |
| storage condition desiccated | storage condition desiccated | storage condition desiccated | storage condition desiccated |
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




