Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C22H24N2O2 · 2HCl
CAS Number:
Molecular Weight:
421.36
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12161501
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
To inquire about this product SML0372, please contact your local Merck office or dealer. Contact Technical Service
Quality Segment
assay
≥98% (HPLC)
form
powder
storage condition
desiccated
color
white to beige
solubility
H2O: 5 mg/mL (clear solution)
storage temp.
room temp
SMILES string
Cl.Cl.COc1ccc(cc1)-c2cc(C(O)C3CCCCN3)c4ccccc4n2
InChI
1S/C22H24N2O2.2ClH/c1-26-16-11-9-15(10-12-16)21-14-18(17-6-2-3-7-19(17)24-21)22(25)20-8-4-5-13-23-20;;/h2-3,6-7,9-12,14,20,22-23,25H,4-5,8,13H2,1H3;2*1H
InChI key
WGWOQHWZXLGPNG-UHFFFAOYSA-N
Biochem/physiol Actions
NSC23925 is a specific Pgp1 inhibitor.
NSC23925 is a specific Pgp1 inhibitor. It reverses MDR1 (Pgp1) but does not inhibit MRP or BCRP-mediated MDR. NSC23925 itself is not a substrate of Pgp1 but stimulates ATPase activity of Pgp, directly inhibiting the function of Pgp1 in a dose-dependent manner without altering the total expression level of Pgp1.
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
