Documents
SIAL0165
Sigma-Aldrich
Sigma® cell culture dish
diam. × H 35 mm × 10 mm, tissue-culture treated, sterile, pack of 25 × 20 ea
About This Item
UNSPSC Code:
41121812
NACRES:
NB.22
sterility
sterile
packaging
pack of 25 × 20 ea
diam. × H
35 mm × 10 mm
surface area
8 cm2
binding type
Tissue Culture (TC)-treated surface
General description
  • Manufactured from optically-clear virgin polystyrene
  • Treated for optimal cell attachment
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Have stacking beads to aid in handling
  • Lid supplied with vents to provide consistent gas exchange
Legal Information
Sigma is a registered trademark of Sigma-Aldrich Co. LLC
Visit the Document Library
Protocols
Universal Transfection Reagent enables efficient nucleic acid delivery into various cells, compatible with different cell culture conditions.
Calcium phosphate transfection is a common method for the introduction of DNA into eukaryotic cells. This protocol can be optimized for use with a wide variety of cell types.