Skip to Content
Merck
All Photos(1)

Key Documents

SAE0081

Sigma-Aldrich

Lipodisq® Styrene:Maleic Anhydride Copolymer 4:1

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 7,360.00

CZK 7,360.00


Please contact Customer Service for Availability


Select a Size

Change View
100 MG
CZK 7,360.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

CZK 7,360.00


Please contact Customer Service for Availability

General description

This product is the anhydride form of the SMA Copolymer 4:1.

Application

Lipodisq reagents are novel polymers derived from styrene and maleic acid. Lipodisq polymers are capable of forming nanosized lipid-based discoidal particles capable of incorporating membrane proteins. The Lipodisq technology can solubilize commonly used lipids such as dimyristoylphosphatidylcholine (DMPC) without the use of detergents. Lipodisqs typically have a diameter of 9-10 nm at pH 7.4.

The Lipodisq technology can solubilize commonly used lipids such as dimyristoylphosphatidylcholine (DMPC) without the use of detergents.

Functional pH range: 8.5 - 10.5

Legal Information

Lipodisq is a registered trademark of Malvern Cosmeceutics Limited

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Francisco Bada Juarez et al.
Chemistry and physics of lipids, 221, 167-175 (2019-04-04)
Nanoparticles assembled with poly(styrene-maleic acid) copolymers, identified in the literature as Lipodisq, SMALPs or Native Nanodisc, are routinely used as membrane mimetics to stabilise protein structures in their native conformation. To date, transmembrane proteins of varying complexity (up to 8

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service