Skip to Content
Merck
All Photos(1)

Documents

SAB5300423

Sigma-Aldrich

Monoclonal Anti-MKI67 antibody produced in mouse

clone 4A1, ascites fluid

Synonym(s):

KIA, Ki-67, MKI67

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

4A1, monoclonal

mol wt

358 kDa

species reactivity

human

technique(s)

direct ELISA: 1:10,000
immunohistochemistry: 1:200-1:1,000
western blot: 1:500-1:2,000

isotype

IgG2b

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MKI67(4288)

Immunogen

Synthetic peptide corresponding to aa (CEDLAGFKELFQTPG) of human KI67, conjugated to KLH.
Mouse monoclonal antibody raised against KI67

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anqi Zeng et al.
Oncology reports, 45(2), 717-727 (2021-01-09)
Breast cancer which is the most common type of diagnosed cancer among women worldwide possesses metastatic potential, multi‑drug resistance, and high mortality. The NF‑κB signaling pathway has been revealed to be abnormally activated in breast cancer cells and closely associated
Francesco Mascadri et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 69(10), 659-667 (2021-09-21)
Antigen-bearing proteins become progressively unavailable to immunodetection after prolonged storage of routine sections, exposed to a variety of agents, such as moisture, oxygen, and temperature. By proteomic analysis, the antigens are retained in the sections and definitely in the tissue
Cunjie Chang et al.
Nature communications, 13(1), 1363-1363 (2022-03-18)
Deregulation of alternative splicing is implicated as a relevant source of molecular heterogeneity in cancer. However, the targets and intrinsic mechanisms of splicing in hepatocarcinogenesis are largely unknown. Here, we report a functional impact of a Splicing Regulatory Glutamine/Lysine-Rich Protein
Zhi Teng et al.
Oncology reports, 43(2), 503-515 (2020-01-03)
Acute lymphoblastic leukemia (ALL) is a malignant hematological disease. Tanshinone IIA (Tan IIA) has antitumor activity in vitro and in vivo. The aim of the present study was to investigate the effects of Tan IIA in combination with imatinib (IM) on the proliferation, apoptosis, migration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service