Skip to Content
Merck
All Photos(1)

Documents

SAB4702141

Monoclonal Anti-CD1a APC produced in mouse

new

Synonym(s):

FCB6, HTA1, Leu-6, T6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

Allophycocyanin conjugate

Quality Level

antibody product type

primary antibodies

clone

SK9, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG2bκ

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD1A(909)

General description

CD1a, together with CD1b and c, belongs to group 1 of CD1 glycoproteins. These proteins serve as antigen-presenting molecules for a subset of T cells that responds to specific lipids and glycolipids found in the cell walls of bacterial pathogens or self-glycolipid antigens such as gangliosides, and they have also roles in antiviral immunity. Unlike CD1b, CD1a is excluded from late endosomal compartments and instead traffics independently in the recycling pathway of the early endocytic system, and CD1a antigen presentation is independent upon vesicular acidification.

Specificity

The mouse monoclonal antibody SK9 recognizes an extracellular epitope of CD1a (T6), a 49 kDa polypeptide associated with beta2-microglobulin expressed on cortical thymocytes (strongly), Langerhans cells, dendritic cells and some T cell leukemias and lymphomas.

Immunogen

Human thymocytes

Physical form

Stabilizing phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified antibody is conjugated with activated allophycocyanin (APC) under optimum conditions and unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.

Storage and Stability

Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service