Skip to Content
Merck
All Photos(1)

Documents

SAB4702036

Monoclonal Anti-CD142 PE produced in mouse

new

Synonym(s):

F3, TF, TFA, coagulation factor III, tissue factor, tissue thromboplastin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

phycoerythrin (R-PE) conjugate

Quality Level

antibody product type

primary antibodies

clone

HTF-1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG1κ

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... F3(2152)

General description

CD142, also known as coagulation factor III, tissue thromboplastin, and tissue factor. It is a transmembrane glycoprotein, which enables cells to initiate the blood coagulation cascades, and functions as the high-affinity receptor for the coagulation factor VII. The resulting complex provides a catalytic event that is responsible for initiation of the coagulation protease cascades by specific limited proteolysis. Unlike the other cofactors of these protease cascades, which circulate as nonfunctional precursors, this factor is a potent initiator that is fully functional when expressed on cell surfaces. It is the only one factor in the coagulation pathway for which a congenital deficiency has not been described.

Specificity

The mouse monoclonal antibody HTF-1, also known as HTF1-7B8, recognizes an extracellular epitope of CD142 (tissue factor, coagulation factor III), a type I glycoprotein expressed on endothelial cells, monocytes, macrophages, and platelets upon induction by inflammatory mediators, and expressed constitutively by some tumors, the vasculature, placenta, kidney, and central nervous system.

Immunogen

Human brain tissue factor (CD142)

Physical form

Stabilizing phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified antibody is conjugated with R-phycoerythrin (PE) under optimum conditions. Unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.

Storage and Stability

Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service