Skip to Content
Merck
All Photos(1)

Key Documents

SAB4500979

Sigma-Aldrich

Anti-MED13L antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Mediator complex subunit 13-like, Mediator of RNA polymerase II transcription subunit 13-like, Thyroid hormone receptor-associated protein 2, Thyroid hormone receptor-associated protein complex 240 kDa component-like

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 9,230.00

CZK 9,230.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
CZK 9,230.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CZK 9,230.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 242 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MED13L(23389)

General description

Anti-MED13L Antibody detects endogenous levels of total MED13L protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human MED13L.

Immunogen Range: 449-498

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service