Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500131

Sigma-Aldrich

Anti-TRXR2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Selenoprotein Z, TR-β, Thioredoxin reductase 2, Thioredoxin reductase TR3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 56 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TXNRD2(10587)

General description

Anti-TRXR2 Antibody detects endogenous levels of total TRXR2 protein.
Thioredoxin reductase 2 (TRXR2) is mitochondrial protein-disulfide oxidoreductase, encoded by the gene mapped to human chromosome 22q11.21. TRXR2 is one of the mammalian isoforms of thioredoxin reductase (TrxR).

Immunogen

The antiserum was produced against synthesized peptide derived from human TRXR2.

Immunogen Range: 471-520

Biochem/physiol Actions

Thioredoxin reductase 2 (TRXR2) regulates cell survival during mammalian embryonic development. In addition, it also regulates mitochondria-dependent apoptosis. TRXR2 also controls protein tyrosine phosphorylation and cell proliferation. The protein expression is associated with development of glioma. In humans, this enzyme stimulates the apoptosis in HeLa cells. Mutation in the gene might increase the risk of susceptibility to myocardial infarction (MI) in type 2 diabetes mellitus (T2DM) patients.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitochondrial thioredoxin system effects of TrxR2 overexpression on redox balance, cell growth, and apoptosis.
Patenaude A, et al.
The Journal of Biological Chemistry, 279(26), 27302-27314 (2004)
Association of thioredoxin reductase 2 (TXNRD2) gene polymorphisms with myocardial infarction in Slovene patients with type 2 diabetes mellitus
Kariz S, et al.
Diabetes Research and Clinical Practice, 108(2), 323-328 (2015)
The Genes Associated with RNA Editing and Glioma on the Chromosome 22q.
Wang G and Tian Y
Advances in Medical Sciences, 2(1) (2014)
Involvements of mitochondrial thioredoxin reductase (TrxR2) in cell proliferation
Kim MR, et al.
Biochemical and Biophysical Research Communications, 304(1), 119-124 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service