Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300160

Sigma-Aldrich

Anti-phospho-BCR (pTyr177) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ALL antibody produced in rabbit, Anti-BCR-ABL1 antibody produced in rabbit, Anti-BCR1 antibody produced in rabbit, Anti-CML antibody produced in rabbit, Anti-breakpoint cluster region antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 9,300.00

CZK 9,300.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
CZK 9,300.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

CZK 9,300.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~210 kDa

species reactivity

mouse, human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(P-F-YP-V-N)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr177)

Gene Information

human ... BCR(613)

Immunogen

Peptide sequence around phosphorylation site of tyrosine 177 (P-F-Y(p)-V-N), according to the protein BCR.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

GTPase-activating protein for RAC1 and CDC42. Promotes the exchange of RAC or CDC42-bound GDP by GTP, thereby activating them. Displays serine/threonine kinase activity.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuki Tanaka et al.
Journal of immunology (Baltimore, Md. : 1950), 201(8), 2256-2263 (2018-09-12)
We recently reported that NF-κB-mediated inflammation caused by breakpoint cluster region (BCR) is dependent on the α subunit of casein kinase II (CK2α) complex. In the current study, we demonstrate that presenilin 1 (Psen1), which is a catalytic component of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service