SAB4200287
Anti-Actin, Muscle specific antibody, Mouse monoclonal
clone HHF-35, purified from hybridoma cell culture
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified from hybridoma cell culture
antibody product type
primary antibodies
clone
HHF-35, monoclonal
form
buffered aqueous solution
mol wt
antigen ~42 kDa
species reactivity
rat, bovine, canine, chicken, human, mouse, hamster, monkey
concentration
~1.0 mg/mL
technique(s)
immunocytochemistry: 2-4 μg/mL using A10 cells
immunohistochemistry: suitable
western blot: 0.1-0.2 μg/mL using A10 cell extracts
isotype
IgG1
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... ACTB(60)
General description
Immunogen
Application
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service