Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200075

Sigma-Aldrich

Anti-VAC14 (C-terminal) antibody produced in rabbit

~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ArPIKfyve, Anti-TAX1-binding protein 2, Anti-TAX1BP2, Anti-TAX1BP2 TAX-reactive protein x, Anti-TRX

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
CZK 13,300.00

CZK 13,300.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
CZK 13,300.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CZK 13,300.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~82 kDa

species reactivity

rat, mouse, human

packaging

antibody small pack of 25 μL

concentration

~1.5 mg/mL

technique(s)

immunoprecipitation (IP): 10-15 μg using A431 cell lysates
indirect immunofluorescence: 8-16 μg/mL using HeLa cells
western blot: 1-2 μg/mL using extracts of mouse brain (S1 fraction) or rat brain (S1 fraction)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VAC14(55697)
rat ... Vac14(307842)

General description

VAC14 gene is mapped to human chromosome 16q22.1-q22.2. VAC14 is a scaffold protein and is a component of the PIKfyve protein kinase complex.

Specificity

Anti-VAC14 (C-terminal) specifically recognizes human, mouse, and rat VAC14.

Application

Anti-VAC14 (C-terminal) antibody produced in rabbit has been used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

Mutation in VAC14 gene leads to the development of retinitis pigmentosa (RP).
VAC14 is a regulator of PIP5K3/PIKfyve, a dual specificity kinase.
VAC14 phosphorylates PtdIns(3)P to generate the housekeeping phospholipid PtdIns(3,5)P2. It associates with and upregulate PIKfyve phosphoinositide-5-kinase activity. VAC14-PIP5K3- PtdIns(3,5)P2 pathway is thought to be physiologically linked to insulin activation of glucose transport into the cell. Knockout of the VAC14 gene in mice results in massive neurodegeneration and particularly affected are neurons in the midbrain and of the peripheral sensory system.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PIKfyve: Partners, significance, debates and paradoxes
Shisheva A
Cell Biology International, 32(6), 591-604 (2008)
Gholson J Lyon et al.
Cold Spring Harbor molecular case studies, 5(6) (2019-08-08)
Whole-exome sequencing was used to identify the genetic etiology of a rapidly progressing neurological disease present in two of six siblings with early childhood onset of severe progressive spastic paraparesis and learning disabilities. A homozygous mutation (c.2005G>T, p, V669L) was
ArPIKfyve homomeric and heteromeric interactions scaffold PIKfyve and Sac3 in a complex to promote PIKfyve activity and functionality
Sbrissa D, et al.
Journal of Molecular Biology, 384(4), 766-779 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service