SAB3500740
Anti-FCHSD1 antibody produced in chicken
affinity isolated antibody
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
chicken
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
mol wt
76 kDa
species reactivity
human, mouse
concentration
1 mg/mL
technique(s)
ELISA: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... FCHSD1(89848)
Immunogen
Antibody was raised against a 15 amino acid peptide near the carboxy terminus of human FCHSD1.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Linkage
The action of this antibody can be blocked using blocking peptide SBP3500740.
Physical form
Supplied at 1 mg/mL in PBS with 0.02% sodium azide.
Not finding the right product?
Try our Product Selector Tool.
related product
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service