Skip to Content
Merck
All Photos(2)

Key Documents

SAB3500327

Sigma-Aldrich

Anti-Smac antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-DIABLO

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 12,900.00

CZK 12,900.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
CZK 12,900.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CZK 12,900.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 26 kDa

species reactivity

rat, human, mouse

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Diablo(66593)

Immunogen

Smac antibody was raised against a peptide corresponding to amino acids 225 to 239 of human Smac/DIABLO .

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500327.

Physical form

Supplied in PBS with 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Habich et al.
Cell reports, 26(3), 759-774 (2019-01-17)
Disulfide formation in the mitochondrial intermembrane space (IMS) is an essential process. It is catalyzed by the disulfide relay machinery, which couples substrate import and oxidation. The machinery relies on the oxidoreductase and chaperone CHCHD4-Mia40. Here, we report on the
Karthik Mohanraj et al.
EMBO molecular medicine, 11(5) (2019-03-20)
Nuclear and mitochondrial genome mutations lead to various mitochondrial diseases, many of which affect the mitochondrial respiratory chain. The proteome of the intermembrane space (IMS) of mitochondria consists of several important assembly factors that participate in the biogenesis of mitochondrial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service