Skip to Content
Merck
All Photos(2)

Key Documents

SAB2104018

Sigma-Aldrich

Anti-NKX6-1, (N-terminal) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-NKX6A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
horse, human, bovine, rat, mouse, pig, rabbit, dog
citations:
technique(s):
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

38 kDa

species reactivity

horse, human, bovine, rat, mouse, pig, rabbit, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NKX6-1(4825)

Immunogen

Synthetic peptide directed towards the N terminal region of human NKX6-1

Biochem/physiol Actions

NKX6-1 is the transcription factor which binds to specific A/T-rich DNA sequences in the promoter regions of a number of genes. NKX6-1 is involved in transcriptional regulation in islet beta cells. Binds to the insulin promoter and is involved in regulation of the insulin gene. Together with NKX2-2 and IRX3, NKX6-1 acts to restrict the generation of motor neurons to the appropriate region of the neural tube. NKX6-1 belongs to the class II proteins of neuronal progenitor factors, which are induced by SHH signals.In the pancreas, NKX6.1 is required for the development of beta cells and is a potent bifunctional transcription regulator that binds to AT-rich sequences within the promoter region of target genes Iype et al. (2004) [PubMed 15056733].[supplied by OMIM]. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-300 U66797.1 1-300 301-307 AC096766.3 119394-119400 c 308-676 U66797.1 308-676 677-849 U66798.1 7-179 850-1116 U66799.1 7-273

Sequence

Synthetic peptide located within the following region: MLAVGAMEGTRQSAFLLSSPPLAALHSMAEMKTPLYPAAYPPLPAGPPSS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service