Skip to Content
Merck
All Photos(2)

Key Documents

SAB1409253

Sigma-Aldrich

Monoclonal Anti-INSRR antibody produced in mouse

clone 2C1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

IRR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2C1, monoclonal

form

buffered aqueous solution

mol wt

antigen 37.73 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... INSRR(3645)

General description

The gene INSRR (insulin receptor related receptor) is mapped to human chromosome 1q21-q23. It belongs to the insulin receptor family of proteins. INSRR is mainly present in organs which are associated with acid/base generation. The protein has two N-terminal leucine-rich repeat domains, a cysteine-rich C domain and three C-terminal fibronectin type-III repeats.

Immunogen

INSRR (NP_055030, 651 a.a. ~ 760 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
LYLNDYCHRGLRLPTSNNDPRFDGEDGDPEAEMESDCCPCQHPPPGQVLPPLEAQEASFQKKFENFLHNAITIPISPWKVTSINKSPQRDSGRHRRAAGPLRLGGNSSDF

Biochem/physiol Actions

INSRR (insulin receptor related receptor) exhibits pH-sensing property and is a sensor for mild alkaline extracellular medium. In mice, it is associated with excessive excretion of base by the kidneys.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The protein tyrosine kinase family of the human genome.
Robinson DR, et al.
Oncogene, 19, 5548-5557 (2000)
Mapping of alkali-sensing sites of the insulin receptor-related receptor. The role of L2 and fibronectin domains.
Deyev IE
Biochimie, 111, 1-9 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service