Skip to Content
Merck
All Photos(3)

Key Documents

SAB1405970

Sigma-Aldrich

Anti-HSPA2 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

HSP70-2, HSP70-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~70 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSPA2(3306)

Immunogen

HSPA2 (NP_068814.2, 1 a.a. ~ 639 a.a) full-length human protein.

Sequence
MSARGPAIGIDLGTTYSCVGVFQHGKVEIIANDQGNRTTPSYVAFTDTERLIGDAAKNQVAMNPTNTIFDAKRLIGRKFEDATVQSDMKHWPFRVVSEGGKPKVQVEYKGETKTFFPEEISSMVLTKMKEIAEAYLGGKVHSAVITVPAYFNDSQRQATKDAGTITGLNVLRIINEPTAAAIAYGLDKKGCAGGEKNVLIFDLGGGTFDVSILTIEDGIFEVKSTAGDTHLGGEDFDNRMVSHLAEEFKRKHKKDIGPNKRAVRRLRTACERAKRTLSSSTQASIEIDSLYEGVDFYTSITRARFEELNADLFRGTLEPVEKALRDAKLDKGQIQEIVLVGGSTRIPKIQKLLQDFFNGKELNKSINPDEAVAYGAAVQAAILIGDKSENVQDLLLLDVTPLSLGIETAGGVMTPLIKRNTTIPTKQTQTFTTYSDNQSSVLVQVYEGERAMTKDNNLLGKFDLTGIPPAPRGVPQIEVTFDIDANGILNVTAADKSTGKENKITITNDKGRLSKDDIDRMVQEAERYKSEDEANRDRVAAKNALESYTYNIKQTVEDEKLRGKISEQDKNKILDKCQEVINWLDRNQMAEKDEYEHKQKELERVCNPIISKLYQGGPGGGSGGGGSGASGGPTIEEVD

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth Bromfield et al.
Molecular human reproduction, 21(10), 755-769 (2015-07-15)
While a large cohort of sperm surface receptors underpin sperm-oocyte adhesion processes, our recent work has revealed that the molecular chaperone Heat Shock Protein A2 (HSPA2) is a key regulator of zona pellucida-receptor complex assembly in our own species. Indeed
Dorota Scieglinska et al.
Cell stress & chaperones, 20(2), 221-235 (2014-10-26)
The HSPA2 gene is a poorly characterized member of the HSPA (HSP70) family. HSPA2 was originally described as testis-specific and expressed at the highest level in pachytene spermatocytes of rodents, the expression of which is not induced by heat shock.
Kate A Redgrove et al.
Molecular human reproduction, 19(3), 120-135 (2012-12-19)
A unique characteristic of mammalian spermatozoa is that, upon ejaculation, they are unable to recognize and bind to an ovulated oocyte. These functional attributes are only realized following the cells' ascent of the female reproductive tract whereupon they undergo a
Dorota Scieglinska et al.
International journal of molecular sciences, 21(12) (2020-06-21)
Heat shock proteins (HSPs) constitute a major part of the molecular chaperone system and play a fundamental role in cell proteostasis. The HSPA (HSP70) family groups twelve highly homologous HSPA proteins. Certain HSPAs are regarded as important cancer-related proteins, prospective
Elizabeth G Bromfield et al.
Molecular human reproduction, 23(2), 91-105 (2016-12-10)
Does oxidative stress compromise the protein expression of heat shock protein A2 (HSPA2) in the developing germ cells of the mouse testis? Oxidative stress leads to the modification of HSPA2 by the lipid aldehyde 4-hydroxynonenal (4HNE) and initiates its degradation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service