Skip to Content
Merck
All Photos(2)

Key Documents

SAB1403096

Sigma-Aldrich

Monoclonal Anti-SLC27A2 antibody produced in mouse

clone 1F1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

ACSVL1, FACVL1, FATP2, HsT17226, VLACS, VLCS, hFACVL1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 12,800.00

CZK 12,800.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
CZK 12,800.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CZK 12,800.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
immunofluorescence: suitable
indirect ELISA: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC27A2(11001)

General description

The protein encoded by this gene is an isozyme of long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme activates long-chain, branched-chain and very-long-chain fatty acids containing 22 or more carbons to their CoA derivatives. It is expressed primarily in liver and kidney, and is present in both endoplasmic reticulum and peroxisomes, but not in mitochondria. Its decreased peroxisomal enzyme activity is in part responsible for the biochemical pathology in X-linked adrenoleukodystrophy. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

SLC27A2 (NP_003636, 25 a.a. ~ 123 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
YFFQDIGYFLKVAAVGRRVRSYGQRRPARTILRAFLEKARQTPHKPFLLFRDETLTYAQVDRRSNQVARALHDHLGLRQGDCVALLMGNEPAYVWLWLG

Biochem/physiol Actions

Solute carrier family 27 member 2 (SLC27A2) has a role in fatty acid transport. It also mediates the activation of fatty acids having a chain length of 22 or more carbons.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S J Steinberg et al.
Molecular genetics and metabolism, 68(1), 32-42 (1999-09-10)
Activation of fatty acids, catalyzed by acyl-coenzyme A (acyl-CoA) synthetases, is required for their subsequent metabolism. Peroxisomes and microsomes contain very-long-chain acyl-CoA synthetases (VLCSs) capable of activating fatty acids with a chain length of 22 or more carbons. Decreased peroxisomal
Elaina M Melton et al.
The Journal of biological chemistry, 286(35), 30670-30679 (2011-07-20)
The trafficking of fatty acids across the membrane and into downstream metabolic pathways requires their activation to CoA thioesters. Members of the fatty acid transport protein/very long chain acyl-CoA synthetase (FATP/Acsvl) family are emerging as key players in the trafficking

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service