Skip to Content
Merck
All Photos(2)

Key Documents

SAB1403056

Sigma-Aldrich

Monoclonal Anti-FLOT1 antibody produced in mouse

clone 4A1, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

4A1, monoclonal

mol wt

antigen ~37.11 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... FLOT1(10211)

General description

Flotillin 1 (FLOT1) is an integral membrane protein and a component of lipid rafts. This estrogen responsive gene is localized on human chromosome 6p21.33.

Immunogen

FLOT1 (NP_005794.1, 126 a.a. ~ 225 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
KFSEQVFKVASSDLVNMGISVVSYTLKDIHDDQDYLHSLGKARTAQVQKDARIGEAEAKRDAGIREAKAKQEKVSAQYLSEIEMAKAQRDYELKKAAYDI

Biochem/physiol Actions

Flotillin 1 (FLOT1) has a role in endocytosis. It also functions in growth factor signaling and activates mitogen-activated protein kinase signaling. The protein has been associated with tumor progression and is expressed in many cancers.

Physical form

Clear solution

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integrative analysis for finding genes and networks involved in diabetes and other complex diseases
Regine Bergholdt
Genome Biology (2007)
Upregulation of flotillin-1 promotes invasion and metastasis by activating TGF-? signaling in nasopharyngeal carcinoma.
Cao S
Oncotarget (2016)
Flotillin-1 protein is upregulated in human endometrial cancer and localization shifts from epithelial to stromal with increasing tumor grade.
Winship AL
Cancer Investigation (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service