Skip to Content
Merck
All Photos(1)

Documents

SAB1306172

Sigma-Aldrich

ANTI-HIF1ALPHA(C-TERMINAL) antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

BHLHE78, HIF1A, Hypoxia-inducible factor 1-alpha, MOP1, PASD8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

92670 Da

species reactivity

human, mouse

technique(s)

immunofluorescence: 1:10-1:50
immunohistochemistry: 1:10-1:50
western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HIF1A(3091)

General description

Hypoxia-inducible factor-1 (HIF1) is a transcription factor expressed in mammalian cells when cultured under reduced oxygen tension. HIF1 is a heterodimer composed of an α subunit and a β subunit. The β subunit has been identified as the aryl hydrocarbon receptor nuclear translocator (ARNT). This gene encodes the alpha subunit of HIF-1. HIF-1α contains two transactivation domains. The gene encoding it is localized on human chromosome 14q23.2.

Biochem/physiol Actions

Hypoxia-inducible factor-1 (HIF1) plays an essential role in cellular and systemic homeostatic responses to hypoxia. During hypoxia, the two subunits of this factor undergo post-translational modifications, which in turn promote transactivation. Overexpression of a natural antisense transcript (aHIF) of this gene has been shown to be associated with nonpapillary renal carcinomas. HIF-1α is a master regulator of the hypoxic response, and its proangiogenic activities include, but are not limited to, regulation of vascular endothelial growth factor (VEGF).

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiple Gastrointestinal Stromal and Other Tumors Caused by Platelet-Derived Growth Factor Receptor a Gene Mutations: A Case Associated with a Germline V561D Defect
Barbara Pasini, et al.
The Journal of Clinical Endocrinology and Metabolism (2007)
Hypoxia-Inducible Factor (HIF)-1 Regulatory Pathway and its Potential for Therapeutic Intervention in Malignancy and Ischemia
The Yale Journal of Biology and Medicine (2007)
Regulation of Hypoxia-Inducible Factor 1a Expression and Function by the Mammalian Target of Rapamycin
Christine C, et al.
Molecular and Cellular Biology (2002)
Vascular endothelial growth factor (VEGF) regulation by hypoxia inducible factor-1 alpha (HIF1A) starts and peaks during endometrial breakdown, not repair, in a mouse menstrual-like model
Xihua Chen, et al.
Human Reproduction (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service