Skip to Content
Merck
All Photos(1)

Key Documents

SAB1305744

Sigma-Aldrich

ANTI-S. AUREUS DELTA HEMOLYSIN antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Delta-hemolysin, Delta-toxin, hld

Sign Into View Organizational & Contract Pricing

Select a Size

400 μL
CZK 11,700.00

CZK 11,700.00


Please contact Customer Service for Availability


Select a Size

Change View
400 μL
CZK 11,700.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CZK 11,700.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

2979 Da

species reactivity

rat

technique(s)

western blot: 1:1000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Woodvine Otieno et al.
Journal of microbiology and biotechnology, 31(9), 1200-1209 (2021-07-29)
Sepsis is an acute inflammatory response that leads to life-threatening complications if not quickly and adequately treated. Cytolysin, hemolysin, and pneumolysin are toxins produced by gram-positive bacteria and are responsible for resistance to antimicrobial drugs, cause virulence and lead to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service