Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300591

Sigma-Aldrich

Anti-DHX37 (center) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-DDX37, Anti-DEAH box protein 37, Anti-DHX37, Anti-KIAA1517, Anti-Probable ATP-dependent RNA helicase DHX37

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 8,030.00

CZK 8,030.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1524


Select a Size

Change View
100 μG
CZK 8,030.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CZK 8,030.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1524

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DHX37(57647)

Related Categories

General description

DHX37 is a DEAD box protein. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division.

Immunogen

DHX37 (NP_116045, 488-527)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the center region of human DHX37.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service