Skip to Content
Merck

S9447

Monoclonal Anti-α-SNAP antibody produced in mouse

~1 mg/mL, clone 4E4, purified immunoglobulin, buffered aqueous glycerol solution

Synonym(s):

Anti-N-ethylmaleimide sensitive factor attachment protein, alpha, Anti-SNAPA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
4E4, monoclonal
Application:
IP, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4E4, monoclonal

form

buffered aqueous glycerol solution

mol wt

antigen ~36 kDa

species reactivity

bovine, human, rat

concentration

~1 mg/mL

technique(s)

immunoprecipitation (IP): suitable, western blot: 1:5,000 using rat brain and kidney

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NAPA(8775)
rat ... Napa(140673)

Immunogen

recombinant human α-SNAP. The epitope resides within the N-terminal domain between amino acids 1 and 109.

Physical form

Solution in 20 mM sodium phosphate, 150 mM sodium chloride, 50% glycerol, and 3 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library