Skip to Content
Merck

S9276

Sodium bicarbonate-14C

buffered aqueous solution, extent of labeling: 2-10 mCi per mmol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
NaH14CO3
CAS Number:
Molecular Weight:
84.01
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

buffered aqueous solution

extent of labeling

2-10 mCi per mmol

mass shift

M+2

storage temp.

2-8°C

SMILES string

[H]O[14C](=O)O[Na]

Packaging

serum bottle

Physical form

Aqueous solution, pH 9.5


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library