Skip to Content
Merck

Skip To

S9059

Streptavidin−Gold from Streptomyces avidinii

~10 nm nominal (colloidal gold), buffered aqueous glycerol solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
0.25 mL
Please contact Customer Service for Availability
CZK 4,750.00
1 mL
Please contact Customer Service for Availability
CZK 12,200.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
gold conjugate
Species reactivity:
-
Application:
Citations:
1

CZK 4,750.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

gold conjugate

Quality Segment

form

buffered aqueous glycerol solution

concentration

~2.5 A520 units/mL

nominal

~10 nm (colloidal gold)

mean particle size

8-12 nm (monodisperse)

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Strepavidin-collodial gold conjugate binds to molecules, such as primary antibodies, that have been previously conjugated with biotin.
Streptavidin is a protein produced by the bacterium Streptomyces avidinii. It has four binding sites for biotin as does avidin. Both have been used extensively as probes in immunochemical systems, conjugated to antibodies, enzymes, fluorochromes and gold. Avidin is highly basic (pI ~ 10.5), compared to streptavidin (pI ~ 5-6). Avidin has a slightly higher binding capacity for biotin than streptavidin does, but it has glycosylated side chains which give higher backgrounds in blotting techniques than does streptavidin, which is not a glycoprotein. Streptavidin is more resistant than avidin to dissociation into subunits by guanidinium chloride. Streptavidin may be conjugated to colloidal gold.

Application

For detection of biotinylated compounds.
Strepavidin-collodial gold conjugate is a component of streptavidin/colloidal gold-biotin detection systems use in immunoblotting, light microscopy, and immunoelectron microscopy applications. It is commonly used in postembedding immunocytochemistry.

Physical form

Suspension in 0.01 M phosphate buffered saline, pH 7.2, containing 1% BSA, 20% glycerol, and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S6402S9415S2890
conjugate

gold conjugate

conjugate

CY3 conjugate

conjugate

-

conjugate

alkaline phosphatase conjugate

form

buffered aqueous glycerol solution

form

buffered aqueous solution

form

-

form

lyophilized powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

concentration

~2.5 A520 units/mL

concentration

-

concentration

-

concentration

-

target post-translational modification

unmodified

target post-translational modification

-

target post-translational modification

-

target post-translational modification

-

nominal

~10 nm (colloidal gold)

nominal

-

nominal

-

nominal

-


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. Can you provide details on how streptavidin is conjugated to gold nanoparticles? Specifically, I'd like to know if there is any covalent binding involved or if it's a passive binding process.

    1 answer
    1. Product S9059 was manufactured using passive conjugation of the STP with 10 nm gold colloid. It does not involve covalent or linker chemistry. The protein binds to the gold colloid through amino acid residues, including charge attraction (lysine), hydrophobic attraction (tryptophan), and sulfur bonding (cysteine).

      Helpful?

  2. I want to know the streptavidin concentration in this stock. (e.g. 0.5 mg/mL)

    1 answer
    1. This information is reported as the protein concentration in the lot-specific Certificate of Analysis. A review of lots available in the last 7 years reveal a range of 11 - 11.6 ug/mL. Please see the link below to review a sample or lot-specific Certificate of Analysis:
      https://www.sigmaaldrich.com/US/en/product/sigma/s9059#product-documentation

      Helpful?

Reviews

No rating value

Active Filters