Skip to Content
Merck

S7546

Zorbax® LP 100/40 CN

40 μm mean particle size, average diameter 100 Å

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
41115700
EC Number:
271-537-8
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mean particle size

40 μm

pore size

100 Å average diameter

InChI

1S/C6H12ClNSi.O2Si/c1-9(2,7)6-4-3-5-8;1-3-2/h3-4,6H2,1-2H3;

InChI key

MPHCKNJRPAUOCD-UHFFFAOYSA-N

Application

Cyano-derivatized silica for normal- or reversed-phase separations of slightly polar molecules

Legal Information

Zorbax is a registered trademark of Agilent Technologies, Inc.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library