Skip to Content
Merck

S4687

Anti-SAP-102 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat

technique(s)

immunohistochemistry: suitable, western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

rat ... Dlg3(58948)

Immunogen

synthetic peptide corresponding to amino acids 83-100 of rat SAP-102.

Physical form

Solution in phosphate buffered saline containing 0.05% sodium azide and 1 mg/ml bovine serum albumin


Still not finding the right product?

Try our Product Selector Tool to narrow your options




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library