Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
MDL number:
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 32-35 kDa (doublet)
species reactivity
canine, human
technique(s)
immunoprecipitation (IP): 5-10 μg using 293-T transfected cell extracts, indirect immunofluorescence: 2-4 μg/mL using 293-T cells transfected with human Sprouty 2., western blot: 2-4 μg/mL using cell extracts of MDCK (Madin Darby canine kidney).
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... SPRY2(10253)
Immunogen
synthetic peptide corresponding to amino acids 301-315 of human Sprouty 2, conjugated to KLH via a N-terminal added cysteine residue.
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.