Merck
S1815
SecureSlip glass coverslip
L × W 12 mm × 12 mm
Synonym(s):
cover slips, coverslips, glass cover slips
Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
NACRES:
NB.22
UNSPSC Code:
41122600
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
sterility
(sterile), sterile
packaging
pkg of 15 coverslips per insert, pkg of 5 inserts
manufacturer/tradename
Grace Bio-Labs
L × W
12 mm × 12 mm
General description
Coverslips are affixed to a thin microscopically transparent silicone base which secures the coverslip to culture vessels to prevent movement and cell damage during cultivation and assay procedures. During culture, cells may be imaged without interference from the silicone. Silicone perimeter surrounding each coverslip acts as a hydrophobic barrier to isolate liquids. Silicone backing removes easily (by peeling) for conventional mounting. Available as inserts which facilitate cultivation and batch processing in sets of up to 15 coverslips per plate or as individual silicone backed 12 mm × 12 mm coverslips.
Legal Information
SecureSlip is a trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)