Skip to Content
Merck

S1563

SKF-86466 hydrochloride

solid, ≥98% (HPLC)

Synonym(s):

6-Chloro-2,3,4,5-tetrahydro-3-methyl-1H-3-benzazepine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H14ClN·HCl
CAS Number:
Molecular Weight:
232.15
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

solid

color

white

solubility

H2O: 24 mg/mL

SMILES string

Cl.CN1CCc2cccc(Cl)c2CC1

InChI

1S/C11H14ClN.ClH/c1-13-7-5-9-3-2-4-11(12)10(9)6-8-13;/h2-4H,5-8H2,1H3;1H

InChI key

JKQKVNMNAIFCFS-UHFFFAOYSA-N

Gene Information

Application

SKF 86466 was used to study adrenoceptor signaling in rat limbic seizure models.

Biochem/physiol Actions

SKF 86466 binds specifically to α2-adrenoceptors at pre- and post-junctional sites but was also reported to antagonize α1-aderenoceptors in humans. The structural similarity of receptors enables SKF 86466 binding to 5-HT1A receptors.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library