Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 mL | Please contact Customer Service for Availability | CZK 3,370.00 CZK 2,864.50 |
5 mL | Please contact Customer Service for Availability | CZK 10,700.00 CZK 9,095.00 |
About This Item
NACRES:
NA.71
UNSPSC Code:
12352207
Biological source:
bovine
Origin:
USA origin
Form:
lyophilized powder
CZK 2,864.50
List PriceCZK 3,370.00Save 15%Please contact Customer Service for Availability
biological source
bovine
form
lyophilized powder
origin
USA origin
storage temp.
2-8°C
Application
Adult bovine serum is bactericidal against Helicobacter pylori. However, it can be used effectively to support H. pylori cultures after heat inactivation at 56 ° C for 30 min.
1 of 1
This Item | |||
|---|---|---|---|
| biological source bovine | biological source bovine | biological source bovine | biological source bovine |
| origin USA origin | origin USA origin | origin USA origin | origin USA origin |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

