Skip to Content
Merck
All Photos(1)

Key Documents

RES1457A

SAFC

L-Aspartic acid monosodium salt monohydrate

Synonym(s):

L-Aspartic acid sodium salt monohydrate, (S)-2-Aminobutanedioic acid sodium salt, Sodium L-aspartate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH(NH2)CO2Na · H2O
CAS Number:
Molecular Weight:
173.10
Beilstein:
6101944
EC Number:
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.21

Quality Level

SMILES string

[Na+].[H]O[H].N[C@@H](CC(O)=O)C([O-])=O

InChI

1S/C4H7NO4.Na.H2O/c5-2(4(8)9)1-3(6)7;;/h2H,1,5H2,(H,6,7)(H,8,9);;1H2/q;+1;/p-1/t2-;;/m0../s1

InChI key

PPTHNBYUFXSJPS-JIZZDEOASA-M

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Biochem/physiol Actions

Principal neurotransmitter for fast synaptic excitation.

Packaging

Product is available in the following package size:

RES1457A-A103X 1 KG

RES1457A-A104X 10 KG

RES1457A-A105X 5 KG

RES1457A-A106X 25 KG

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service