Skip to Content
Merck

Skip To

RABTYROE

HRP-Conjugated Phosphotyrosine-specific Antibody Concentrate (Item E)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 μL
Please contact Customer Service for Availability
391,00 CZK

About This Item

NACRES:
NA.32
UNSPSC Code:
12352203
Clone:
-
Species reactivity:
-
Application:
Citations:
-

391,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


antibody product type

primary antibodies

shipped in

wet ice

storage temp.

−20°C

Other Notes

This item is for RAB series CBEL ELISA kits.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RABHRP2RABHRP1SAB2108897
shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

antibody product type

primary antibodies

antibody product type

primary antibodies

antibody product type

primary antibodies

antibody product type

primary antibodies


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters