Skip to Content
Merck

Skip To

RAB1906

Rat IL-17F ELISA

for serum, plasma and cell culture supernatants

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
96 wells
Please contact Customer Service for Availability
21 600,00 CZK

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
rat ... Il17f(301291)
Input:
sample type serum
sample type cell culture supernatant(s)
sample type plasma
Species reactivity:
rat
Storage temp.:
−20°C
Shipped in:
wet ice

21 600,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.41 ng/mL
standard curve range: 0.410-100 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Il17f(301291)

General description

This ELISA antibody pair detects Rat IL-17F

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB1911RAB1908RAB1147
description

for serum, plasma and cell culture supernatants

description

for serum, plasma and cell culture supernatants

description

for serum, plasma and cell culture supernatants

description

for cell culture supernatants, plasma, and serum samples

Gene Information

rat ... Il17f(301291)

Gene Information

rat ... Il9(116558)

Gene Information

rat ... Il22(500836)

Gene Information

rat ... Il18(29197)

species reactivity

rat

species reactivity

rat

species reactivity

rat

species reactivity

rat

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.41 ng/mL
standard curve range: 0.410-100 ng/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.45 ng/mL
standard curve range: 0.410-100 ng/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.06 ng/mL
standard curve range: 0.061-15 ng/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1.6 ng/ml

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

input

sample type cell culture supernatant(s)
sample type serum sample(s)
sample type plasma

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters