Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Please contact Customer Service for Availability | 21 600,00 CZK |
About This Item
sample type cell culture supernatant(s)
sample type serum
21 600,00 CZK
species reactivity
rhesus macaque
technique(s)
ELISA: suitable
input
sample type plasma
sample type cell culture supernatant(s)
sample type serum
assay range
inter-assay cv: <12%
intra-assay cv: <10%
shipped in
wet ice
storage temp.
−20°C
Gene Information
rhesus macaque ... PI3(574237)
General description
Application
Please refer to the attached Protocolfor details.
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description for serum, plasma and cell culture supernatants | description for serum, plasma and cell culture supernatants | description for serum, plasma and cell culture supernatants | description for serum, plasma and cell culture supernatants |
| species reactivity rhesus macaque | species reactivity rhesus macaque | species reactivity rhesus macaque | species reactivity rhesus macaque |
| Gene Information rhesus macaque ... PI3(574237) | Gene Information rhesus macaque ... IGFBP2(696214) | Gene Information rhesus macaque ... CCL4L1(574217) | Gene Information rhesus macaque ... PLG(703891) |
| input sample type plasma | input sample type cell culture supernatant(s) | input sample type plasma | input sample type plasma |
| assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% |
| technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




