Skip to Content
Merck

RAB1625

Human Park7 ELISA

for serum, plasma and cell culture supernatants

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... PARK7(11315)
Input:
sample type plasma
sample type serum
sample type cell culture supernatant(s)
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 32 pg/mL
standard curve range: 32.77-8000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PARK7(11315)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library